Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF765 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191630
Description
ZNF765 Polyclonal specifically detects ZNF765 in Human samples. It is validated for Western Blot.Specifications
ZNF765 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
zinc finger protein 765 | |
Rabbit | |
Affinity purified | |
RUO | |
91661 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_001035275 | |
ZNF765 | |
The immunogen for this antibody is ZNF765. Peptide sequence CHRRLHTGEKPYKCEECDKAFHFKSKLQIHRRIHTGEKPYKCNECGKTFS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Mouse: 92%; Sumatran orangutan: 90%; Green puffer: 90%; Crab-eating macaque: 90%; Largemouth bass: 90%; Tunicate: 90%; Mosquito: 90%; Body louse: 90%; Rat: 90%; Red flour beetle: 90%; Fruit fly: 90%; Western clawed frog: 90%; Zebrafish: 90%; White-tufted-ear marmoset: 90%; Bovine: 90%; Florida lancelet: 85%; Chicken: 84%; Canine: 84%; Yellowfever mosquito: 84%; African malaria mosquito: 83%; Xenopus: 78%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction