Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP3/NALP3 Antibody (Nalpy3-b) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP197601
Description
NLRP3/NALP3 Monoclonal specifically detects NLRP3/NALP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| NLRP3/NALP3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| NLRP3 | |
| Recombinant human NLRP3/NALP3 Antibody (Nalpy3-b) (NACHT-, LRR- and PYD-containing protein 3) (pyrin domain). | |
| 0.05 mg | |
| Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
| 114548 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Nalpy3-b | |
| Unconjugated | |
| AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes human NLRP3/NALP3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction