Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM15B Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen RBM15B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP182412
SDP
View Documents
Novus Biologicals
NBP182412
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

RBM15B Polyclonal specifically detects RBM15B in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

RBM15B
Polyclonal
Rabbit
NP_780611
29890
Synthetic peptide towards Rbm 15b. Peptide sequence SPYEERSRTKGGGQQSERGSDRTPERSRKENHSSEGTKESGSNSLSNSRH.
Primary
Western Blot
Unconjugated
RUO
chromosome 3p21.1 gene sequence, HUMAGCGB, one-twenty two protein 3, RNA binding motif protein 15B, RNA-binding motif protein 15B
RBM15B
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.