Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNCX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19131420UL
Description
UNCX Polyclonal specifically detects UNCX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| UNCX | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_001073930 | |
| UNCX | |
| Synthetic peptide directed towards the C terminal of human UNCX. Peptide sequence AAATEGGGGDCADAGTAGPAPPPPAPSPRPGPRPPSPAEEPATCGVPEPG. | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| homeobox protein unc-4 homolog, homeobox protein Uncx4.1, UNC homeobox | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 340260 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction