Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM144 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TMEM144 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TMEM144 Polyclonal specifically detects TMEM144 in Human samples. It is validated for Western Blot.Specifications
TMEM144 | |
Polyclonal | |
Rabbit | |
NP_060812 | |
55314 | |
Synthetic peptide directed towards the middle region of human TMEM144. Peptide sequence LSTVHHRIVGCSLAVISGVLYGSTFVPIIYIKDHSKRNDSIYAGASQYDL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
transmembrane protein 144 | |
TMEM144 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title