Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUXA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | DUXA |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DUXA Polyclonal specifically detects DUXA in Human samples. It is validated for Western Blot.Specifications
DUXA | |
Polyclonal | |
Rabbit | |
Human | |
double homeobox A, double homeobox protein A | |
DUXA | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
NP_001012747 | |
503835 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title