Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SULT6B1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SULT6B1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19132620
![]() |
Novus Biologicals
NBP19132620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191326
![]() |
Novus Biologicals
NBP191326 |
100 μL |
Each for $487.50
|
|
|||||
Description
SULT6B1 Polyclonal specifically detects SULT6B1 in Human samples. It is validated for Western Blot.Specifications
SULT6B1 | |
Polyclonal | |
Rabbit | |
NP_001027549 | |
391365 | |
Synthetic peptide directed towards the C terminal of human SULT6B1. Peptide sequence FLGFFLTGEQIQTISVQSTFQAMRAKSQDTHGAVGPFLFRKGEVGDWKNL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ST6B1, sulfotransferase 6B1, sulfotransferase family, cytosolic, 6B, member 1 | |
SULT6B1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title