Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEZF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191367
Description
FEZF1 Polyclonal specifically detects FEZF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FEZF1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FEZ, FEZ family zinc finger 1, zinc finger protein 312B, ZNF312B | |
Rabbit | |
Affinity purified | |
RUO | |
389549 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
NP_001019784 | |
FEZF1 | |
Synthetic peptide directed towards the C terminal of human FEZF1. Peptide sequence CPTCGKGFCRNFDLKKHVRKLHDSSLGLARTPAGEPGTEPPPPLPQQPPM. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Rabbit: 92%; Guinea pig: 78%; Equine: 78%. | |
Human, Rat, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction