Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEZF1 Antibody, Novus Biologicals™
SDP

Catalog No. p-7110181 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191367 100 μL
NBP19136720 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP191367 Supplier Novus Biologicals Supplier No. NBP191367
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FEZF1 Polyclonal specifically detects FEZF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FEZF1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001019784
Gene Alias FEZ, FEZ family zinc finger 1, zinc finger protein 312B, ZNF312B
Gene Symbols FEZF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human FEZF1. Peptide sequence CPTCGKGFCRNFDLKKHVRKLHDSSLGLARTPAGEPGTEPPPPLPQQPPM.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389549
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Rabbit: 92%; Guinea pig: 78%; Equine: 78%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Rat, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.