Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF823 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ZNF823 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
ZNF823 Polyclonal specifically detects ZNF823 in Human samples. It is validated for Western Blot.Specifications
| ZNF823 | |
| Polyclonal | |
| Purified | |
| RUO | |
| NP_001073962 | |
| 55552 | |
| Synthetic peptide directed towards the N terminal of human HSZFP36. Peptide sequence GECGEVVLGHSSLNCNIRVDTGHKSCEHQEYGEKPYTHKQRGKAISHQHS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| HSZFP36, ZFP 36 for a zinc finger protein, ZFP36, ZFP-36 for a zinc finger protein, zinc finger protein 823, zinc finger protein ZFP-36 | |
| ZNF823 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title