Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zfp113 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | Zfp113 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Zfp113 Polyclonal specifically detects Zfp113 in Mouse samples. It is validated for Western Blot.Specifications
| Zfp113 | |
| Polyclonal | |
| Rabbit | |
| NP_062721 | |
| 56314 | |
| The specific Immunogen is proprietary information. Peptide sequence PHKCDECSKSFNRTSDLIQHQRIHTGEKPYECSECGKAFSQSAHLIQHQR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| KIAA4229,4732456B05Rik, mKIAA4229, zinc finger protein 113 | |
| ZFP113 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title