Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NLRP4G Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579741
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Mouse Ovary Tissue. IHC: human lung cancer tissue.
The protein encoded by this gene is a member of the nucleotide-binding and leucine-rich repeat receptor (NLR) family, and is predicted to contain an N-terminal pyrin effector domain (PYD), a centrally-located nucleotide-binding and oligomerization domain (NACHT) and C-terminal leucine-rich repeats (LRR). This gene product has a demonstrated role as a negative regulator of autophagy and type I interferon signaling pathways as a result of protein interactions with its NACHT domain. The PYD domain has also been shown to be important in the inhibition of NF-kB (nuclear factor kappa-light-chain-enhancer of activated B cells).
Specifications
NLRP4G | |
Polyclonal | |
Unconjugated | |
Nlrp4g | |
nalp4g; NLR family, pyrin domain containing 4G; Nlrp4g | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
235779 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
0 | |
Nlrp4g | |
A synthetic peptide corresponding to a sequence at the C-terminus of mouse NLRP4G (704-721aa SLQYLDLGCSFLTHSEVK). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction