Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP7 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP219548

Catalog No. NBP219548


Only null left
Add to Cart

Description

Description

NLRP7 Polyclonal antibody specifically detects NLRP7 in Human, Sheep samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

NLRP7
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
CLR19.4, MGC126471, NACHT, leucine rich repeat and PYD containing 7, NACHT, LRR and PYD containing protein 7, NACHT, LRR and PYD domains-containing protein 7, NALP7MGC126470, NLR family, pyrin domain containing 7, NOD12HYDM, Nucleotide-binding oligomerization domain protein 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 7, PAN7, PYPAF3FLJ94610, PYRIN-containing APAF1-like protein 3
Rabbit
112 kDa
0.1 mL
Primary
Human. Species reactivity based on sequence identity: Monkey (88%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.76mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin
Q8WX94
NLRP7
Recombinant protein encompassing a sequence within the center region of human NLRP7. The exact sequence is proprietary.
Affinity Purified
RUO
199713
Human, Sheep
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only