Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP9 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282080MFV500
Description
NLRP9 Polyclonal antibody specifically detects NLRP9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NLRP9 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| CLR19.1, NACHT, leucine rich repeat and PYD containing 9, NACHT, LRR and PYD containing protein 9, NACHT, LRR and PYD domains-containing protein 9, NALP9, NLR family, pyrin domain containing 9, NOD6, nucleotide-binding oligomerization domain protein 6, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 9, PAN12, PYRIN and NACHT-containing protein 12 | |
| NLRP9 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human NLRP9. The immunogen is located within amino acids 190 - 240 of NLRP9. Amino Acid Squence: SRDWPESSEKIEDI | |
| 0.1 mL | |
| Innate Immunity | |
| 338321 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction