Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $426.00
Specifications
Antigen | NLRP9 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 4-6 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224661SS
![]() |
Novus Biologicals
NBP224661SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224661
![]() |
Novus Biologicals
NBP224661 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
NLRP9 Polyclonal specifically detects NLRP9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NLRP9 | |
Western Blot 4-6 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
CLR19.1, NACHT, leucine rich repeat and PYD containing 9, NACHT, LRR and PYD containing protein 9, NACHT, LRR and PYD domains-containing protein 9, NALP9, NLR family, pyrin domain containing 9, NOD6, nucleotide-binding oligomerization domain protein 6, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 9, PAN12, PYRIN and NACHT-containing protein 12 | |
NLRP9 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human and 71% homologous in mouse. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q7RTR0 | |
338321 | |
A portion of amino acids 330-380 of human NALP9 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title