Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLRX1 Monoclonal Antibody (OTI4H8)
GREENER_CHOICE

Catalog No. PIMA527207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527207 Supplier Invitrogen™ Supplier No. MA527207
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NLRX1 Monoclonal Antibody for IHC (P), Western Blot

This gene encodes a member of the NLR family. Alternative splicing has been observed at this gene locus and two transcript variants, encoding distinct isoforms, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NLRX1
Gene Accession No. Q86UT6
Gene Alias BC034204; Caterpiller protein 11.3; CLR11.3; DLNB26; FLJ21478; MGC131937; MGC21025; NLR family member X1; NLRX1; NOD26; NOD5; NOD9; NOD-like receptor X1; nucleotide-binding domain and leucine-rich repeat containing; nucleotide-binding oligomerization domain protein 26; Nucleotide-binding oligomerization domain protein 5; nucleotide-binding oligomerization domain protein 9; nucleotide-binding oligomerization domain, leucine rich repeat containing X1; OTTHUMP00000205768; OTTHUMP00000205769; RGD1311293
Gene Symbols NLRX1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 866-975 of NLRX1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79671
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.