Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NLRX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521018
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521018 Supplier Invitrogen™ Supplier No. PA521018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is rat spleen tissue lysate. PA5-21018 can be used with blocking peptide PEP-1132.

This gene encodes a member of the NLR family. Alternative splicing has been observed at this gene locus and two transcript variants, encoding distinct isoforms, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRX1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NLRX1
Gene Accession No. Q3TL44, Q5FVQ8, Q86UT6
Gene Alias BC034204; Caterpiller protein 11.3; CLR11.3; DLNB26; FLJ21478; MGC131937; MGC21025; NLR family member X1; NLRX1; NOD26; NOD5; NOD9; NOD-like receptor X1; nucleotide-binding domain and leucine-rich repeat containing; nucleotide-binding oligomerization domain protein 26; Nucleotide-binding oligomerization domain protein 5; nucleotide-binding oligomerization domain protein 9; nucleotide-binding oligomerization domain, leucine rich repeat containing X1; OTTHUMP00000205768; OTTHUMP00000205769; RGD1311293
Gene Symbols NLRX1
Host Species Rabbit
Immunogen A 14 amino acid peptide near the amino terminus of human NOD5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270151, 315599, 79671
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.