Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NM23-H1 Antibody (084), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289930PE
Description
NM23-H1 Monoclonal antibody specifically detects NM23-H1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
NM23-H1 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
084 | |
Immunohistochemistry-Paraffin | |
EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein | |
Rabbit immunized with purified, recombinant Human NM23-H1 (Accession#: NP_000260.1, Ala2-Glu152) | |
0.1 mL | |
Lipid and Metabolism | |
4830 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction