Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NM23-H1 Human Recombinant Protein
SDP

Catalog No. NBP2522502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2522502 0.02 mg
NBP25225001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2522502 Supplier Novus Biologicals™ Supplier No. NBP2522500.02MG

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-152 of Human NM23-H1 The Recombinant Human NM23-H1 Protein is derived from E. coli. The Recombinant Human NM23-H1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH7.5) containing 1mM DTT, 10% glycerol
Gene ID (Entrez) 4830
Molecular Weight (g/mol) TMW: 17.1kDa
Quantity 0.02 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein
Common Name NM23-H1
Biological Activity Specific activity is > 1,200 units/mg, and is defined as the amount of enzyme that convert 1.0 umole each of ATP and TDP to ADP and TTP per minute at pH 7.5 at 25C a couple system with PK/LDH.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.