Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NM23-H2/NME2 Antibody (2E3), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260241JF646
This item is not returnable.
View return policy
Description
NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
NM23-H2/NME2 | |
Monoclonal | |
Janelia Fluor 646 | |
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
Recombinant human NM23-H2/NME2 (1-152aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
4831 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
2000 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction