Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NM23-H2/NME2 Antibody (4G7A8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | NM23-H2/NME2 |
---|---|
Clone | 4G7A8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
NM23-H2/NME2 Monoclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NM23-H2/NME2 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
4831 | |
Purified recombinant fragment of human NME2 (AA: FULL(1-152)) expressed in E. Coli. | |
Primary | |
17.3 kDa |
4G7A8 | |
Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
NME2 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title