Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NM23-H2/NME2 Antibody (CPTC-NME2-2), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279873AF488
Description
NM23-H2/NME2 Monoclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NM23-H2/NME2 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NME2 | |
Recombinant full-length human NM23-H2/NME2 protein (Uniprot: P22392) | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
Human | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CPTC-NME2-2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction