Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NM23-H2/NME2, Mouse anti-Human, Clone: CPTC-NME2-2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279738100UG
Description
NM23-H2/NME2 Monoclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
NM23-H2/NME2 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P22392, P22392 | |
NME2 | |
Recombinant full-length human NM23-H2/NME2 protein | |
100 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
4831 | |
Store at 4°C. | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
CPTC-NME2-2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction