Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588384
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588384 Supplier Invitrogen™ Supplier No. PA588384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MARRAGGARM FGSLLLFALL AAGVAPLSWD LPEPRSRASK IRVHSRGNLW ATGHFMGKKS LEPSSPSPLG TAPHTSLRDQ RLQLSHDLLG ILLLKKALGV SLSRPAPQIQ YRRLLVQILQ K; Positive Samples: SKOV3, PC-12, Mouse brain, Mouse kidney.

NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NMB
Gene Accession No. P08949, Q9CR53
Gene Alias 3110023K12Rik; neuromedin B; neuromedin-B; Neuromedin-B-32; neuromedin-beta; NMB; RGD1562710
Gene Symbols NMB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-121 of human NMB (NP_0665632).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4828, 499194, 68039
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.