Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMBR Monoclonal Antibody (466505)
GREENER_CHOICE

Catalog No. PIMA524187
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524187 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524187 Supplier Invitrogen™ Supplier No. MA524187
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody stains human Neuromedin B R/NMBR transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.

NMBR, also known as neuromedin B receptor, a Bombesin Receptor, activates phospholipase and protein kinase C and enhances calcium mobilization. It binds neuromedin B, a potent mitogen and growth factor for normal and neoplastic lung and for gastrointestinal epithelial tissues. Neuromedin B receptor expression has been reported in brain, particularly the olfactory bulb and thalamus, and in colon, liver, placenta, testis, spinal cord, and gastrointestinal tract. In addition, it has been shown to be present in cancer of the lung, ovary, and prostate. ESTs have been isolated from B-cell/lung/testis and brain libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMBR
Applications Flow Cytometry
Classification Monoclonal
Clone 466505
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene NMBR
Gene Accession No. P28336
Gene Alias BB1; BB182387; BB1R; bombesin receptor 1; epididymis secretory sperm binding protein Li 185a; epididymis tissue protein Li 185a; neuromedin B receptor; neuromedin-B receptor; Neuromedin-B-preferring bombesin receptor; NMBR; NMB-R
Gene Symbols NMBR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NMBR Met1-Met390.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4829
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.