Learn More
Invitrogen™ NMBR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102109
Description
Antibody detects endogenous levels of total NMBR.
NMBR, also known as neuromedin B receptor, a Bombesin Receptor, activates phospholipase and protein kinase C and enhances calcium mobilization. It binds neuromedin B, a potent mitogen and growth factor for normal and neoplastic lung and for gastrointestinal epithelial tissues. Neuromedin B receptor expression has been reported in brain, particularly the olfactory bulb and thalamus, and in colon, liver, placenta, testis, spinal cord, and gastrointestinal tract. In addition, it has been shown to be present in cancer of the lung, ovary, and prostate. ESTs have been isolated from B-cell/lung/testis and brain libraries.
Specifications
| NMBR | |
| Polyclonal | |
| Unconjugated | |
| NMBR | |
| BB1; BB182387; BB1R; bombesin receptor 1; epididymis secretory sperm binding protein Li 185a; epididymis tissue protein Li 185a; neuromedin B receptor; neuromedin-B receptor; Neuromedin-B-preferring bombesin receptor; NMBR; NMB-R | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18101, 25264, 4829 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54799, P24053, P28336 | |
| NMBR | |
| A synthesized peptide derived from human NMBR(Accession P28336), corresponding to amino acid residues S229-W279. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.