Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NMD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100291
Description
Antibody detects endogenous levels of total NMD3.
NMD3 is a cytoplasmic factor required for a late cytoplasmic assembly step of the 60S subunit but is not a ribosomal protein. A mutation in NMD3 was found to be lethal in the absence of XRN1, which encodes the major cytoplasmic exoribonuclease responsible for mRNA turnover. The NMD3 protein sequence does not contain readily recognizable motifs of known function. However, NMD3 orthologues display an amino-terminal domain containing four repeats of Cx2C, reminiscent of zinc-binding proteins, implicated in nucleic acid binding or protein oligomerization.
Specifications
NMD3 | |
Polyclonal | |
Unconjugated | |
Nmd3 | |
60S ribosomal export protein NMD3; C87860; CGI-07; hNMD3; Nmd3; NMD3 homolog; NMD3 homolog (S. cerevisiae); NMD3 ribosome export adaptor; RGD1308468 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
310512, 51068, 97112 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96D46, Q99L48 | |
Nmd3 | |
A synthesized peptide derived from human NMD3(Accession Q96D46), corresponding to amino acid residues L385-D435. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction