Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578616 Supplier Invitrogen™ Supplier No. PA578616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse heart Predicted Reactivity: Human (100%), Zebrafish (82%), Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NMD3 is a cytoplasmic factor required for a late cytoplasmic assembly step of the 60S subunit but is not a ribosomal protein. A mutation in NMD3 was found to be lethal in the absence of XRN1, which encodes the major cytoplasmic exoribonuclease responsible for mRNA turnover. The NMD3 protein sequence does not contain readily recognizable motifs of known function. However, NMD3 orthologues display an amino-terminal domain containing four repeats of Cx2C, reminiscent of zinc-binding proteins, implicated in nucleic acid binding or protein oligomerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Nmd3
Gene Accession No. Q96D46, Q99L48
Gene Alias 60S ribosomal export protein NMD3; C87860; CGI-07; hNMD3; Nmd3; NMD3 homolog; NMD3 homolog (S. cerevisiae); NMD3 ribosome export adaptor; RGD1308468
Gene Symbols Nmd3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NMD3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310512, 51068, 97112
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.