Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NMDA
GREENER_CHOICESDP

Catalog No. 0114500 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-145-00 500 mg
01-145-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-145-00 Supplier R&D Systems™ Supplier No. 0114/500

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective NMDA agonist

Prototypic NMDA receptor agonist.

Caged version also available (Cat. No 2224).

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4

Specifications

Quantity 500 mg
Product Line Tocris
Formula Weight 147.13
Purity >99%
Molecular Weight (g/mol) 147.13
Chemical Name or Material NMDA
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.