Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDA R, NR1 Subunit Antibody (1031915), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB10655E
Description
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
NMDA R, NR1 Subunit | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry | |
1031915 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
GluN1, MRD8, NDHMSD, NDHMSR,NMD-R1, NMDA1, NMDAR1, NR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 | |
0.1 mL | |
Cellular Signaling, Neuroscience | |
2902 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction