Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR1 Antibody (S308-48), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259327
Description
NMDAR1 Monoclonal specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NMDAR1 | |
Monoclonal | |
Unconjugated | |
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
GRIN1 | |
Fusion protein amino acids 42-361 (extracellular N-terminus) of rat NR1 | |
100 μg | |
Primary | |
Detects 105kDa. | |
Store at -20C. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S308-48 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1 | |
Mouse | |
Protein G purified | |
RUO | |
2902 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction