Learn More
NMDAR1 (extracellular) Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111814
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: ETEKPRGYQM STRLK(C) Homology: Human, mouse amino acid residues identical
NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
Specifications
NMDAR1 (extracellular) | |
Polyclonal | |
Unconjugated | |
GRIN1 | |
GluN1; GluRdelta1; Glurz1; GluRzeta1; Glutamate [NMDA] receptor subunit zeta-1; Glutamate receptor ionotropic, NMDA 1; Grin1; M100174; MRD8; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2′; NR1N1; pGluN1; Rgsc174; RP11-350O14.1 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
14810, 24408, 2902 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P35438, P35439, Q05586 | |
GRIN1 | |
Peptide corresponding to amino acid residues 385-399 of rat GluN1 (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.