Learn More
Invitrogen™ NMDAR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ OPA104013
Description
This antibody is predicted to react with human based on 100% sequence homology. In Western blot, this antibody detects an ~100 kDa protein representing the NR1 C2 splice variant from rat brain homogenate. It shows no cross-reactivity with the NR1 N1, C1, or C2 splice variants. Reconstitute with 50 μL of PBS.
NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
Specifications
NMDAR1 | |
Polyclonal | |
Unconjugated | |
GRIN1 | |
GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14810, 24408 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
0.5 mg/mL | |
5 mM Ammonium bicarbonate with no preservative | |
P35438, P35439 | |
GRIN1 | |
Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit of rat GRIN1, C2 splice variant insert conjugated to KLH. | |
25 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.