Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR1 Rabbit anti-Human, Mouse, Rat, Clone: 1H1T3, Novus Biologicals™

Rabbit Monoclonal Antibody
$206.50 - $492.50
Specifications
Antigen | NMDAR1 |
---|---|
Clone | 1H1T3 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NMDAR1 Monoclonal antibody specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
NMDAR1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2902 | |
IgG | |
Affinity purified |
1H1T3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1 | |
A synthetic peptide corresponding to a sequence within amino acids 800-900 of human NMDAR1 (Q05586). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title