Learn More
Invitrogen™ NMDAR1 Recombinant Superclonal™ Antibody (1HCLC)
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 710020
Description
This antibody is predicted to react with mouse based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
Specifications
NMDAR1 | |
Recombinant Superclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P35438, P35439, Q05586 | |
GRIN1 | |
Recombinant NMDA Receptor 1 protein corresponding to amino acids 834-938. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1HCLC | |
Unconjugated | |
GRIN1 | |
GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1 | |
Rabbit | |
Protein A | |
RUO | |
14810, 24408, 2902 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.