Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2A Antibody (S327-95), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222405B
Description
NMDAR2A Monoclonal specifically detects NMDAR2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NMDAR2A | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
GRIN2A | |
Fusion protein amino acids 75-325 (extracellular N-terminus) of rat GluN2A/NR2A | |
0.1 mL | |
Primary | |
Detects approx 170kDa. Does not react with NR2B. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S327-95 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
GluN2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1 | |
Mouse | |
Protein G purified | |
Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
2903 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction