Learn More
NMDAR2A/GRIN2A Rabbit anti-Human, Mouse, Rat, Polyclonal, ProteinTech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 285251AP150UL

Description
Aliquoting is unnecessary for -20°C storage.
NMDAR2A/GRIN2A Polyclonal antibody specifically detects NMDAR2A/GRIN2A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NMDAR2A/GRIN2A | |
Polyclonal | |
Unconjugated | |
Grin2a | |
EPND; FESD; GluN2A; GluRepsilon1; glutamate [NMDA] receptor subunit epsilon-1; glutamate ionotropic receptor NMDA type subunit 2A; glutamate receptor; glutamate receptor ionotropic, NMDA 2A; glutamate receptor, ionotropic, NMDA2A (epsilon 1); glutamate receptor, ionotropic, N-methyl D-aspartate 2A; Grin2a; hNR2A; LKS; NMDA Receptor 2A; NMDAR 2A; NMDAR2A; N-methyl D-aspartate receptor subtype 2A; N-methyl-D-aspartate receptor channel, subunit epsilon-1; N-methyl-D-aspartate receptor subunit 2A; NR2A; OTTHUMP00000160135; OTTHUMP00000174531 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14811, 24409, 2903 | |
-20°C | |
Liquid |
Western Blot | |
780 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P35436, Q00959, Q12879 | |
Grin2a | |
NMDAR2A/GRIN2A Fusion Protein Ag29101 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.