Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2B Antibody (S59), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147611IR
Description
NMDAR2B Monoclonal specifically detects NMDAR2B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NMDAR2B | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
Mouse | |
Protein G purified | |
Autophagy, Neurodegeneration, Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
2904 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
S59-36 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q00960 | |
GRIN2B | |
Fusion protein amino acids 20-271 (extracellular N-terminus) of rat NR2B | |
0.1 mL | |
Primary | |
Detects approx 166kDa. No cross-reactivity against NR2A. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction