Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR2B Monoclonal Antibody (S59), FITC
GREENER_CHOICE

Catalog No. PIMA545463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545463 Supplier Invitrogen™ Supplier No. MA545463
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45463 was sufficient for detection of NR2B in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 166kDa. No cross-reactivity against NR2A.

NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer's Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S59
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene GRIN2B
Gene Accession No. Q00960, Q01097, Q13224
Gene Alias AW490526; EIEE27; GluN2B; GluRepsilon2; glutamate [NMDA] receptor subunit epsilon-2; glutamate ionotropic receptor NMDA type subunit 2B; glutamate receptor; glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, NMDA2B; glutamate receptor, ionotropic, NMDA2B (epsilon 2); glutamate receptor, ionotropic, N-methyl D-aspartate 2B; GRIN 2B; GRIN2B; HGNC:4586; hNR 3; hNR3; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2B; MGC142178; MGC142180; MRD6; Nmdar2b; NMDE2; NME2; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 2B; N-methyl-D-aspartate receptor subunit 3; NR2B; NR3
Gene Symbols GRIN2B
Host Species Mouse
Immunogen Fusion protein amino acids 20-271 (extracellular N-terminus) of rat NR2B.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14812, 24410, 2904
Target Species Human, Mouse, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.