Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2B, Mouse anti-Human, Mouse, Rat, Clone: 11D1, Millipore Sigma™

Catalog No. MABN247525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN247525 25 μg
MABN2475100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN247525 Supplier MilliporeSigma Supplier No. MABN247525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NMDAR2B, clone 11D1, Cat. No. MABN2475, is a highly specific mouse monoclonal antibody that targets Glutamate receptor ionotropic, NMDA 2B and has been tested for use in Western Blotting.

Glutamate receptor ionotropic, NMDA2B (UniProt: Q13224; also known GluN2B, Glutamate [NMDA] receptor subunit epsilon-2, N-methyl D-aspartate receptor subtype 2B, NMDAR2B, NR2B, N-methyl-D-aspartate receptor subunit 3, NR3, hNR3) is encoded by the GRIN2B (also known as NMDAR2B) gene (Gene ID: 2904) in human. NMDA receptor complexes are ligand-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. They are involved in long-term potentiation and neuronal plasticity and regulate the number of nerve cells during development. However, over-stimulation NMDA receptors can lead to neuronal degeneration. NMDAR2B is a heterotetrameric, multi-pass membrane protein that is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off in the mature form. The heterotetrameric channel is composed of two zeta subunits and two epsilon subunits, but can also form channels that contain at least one zeta subunit and at least one epsilon subunit. Activation of NMDA 2B channel requires binding of glutamate to the epsilon subunit, glycine binding to the zeta subunit, plus membrane depolarization to eliminate channel inhibition by magnesium ions. NMDA 2B is reported to be phosphorylated at Ser1303 by DAPK1 that enhances its channel activity allowing injurious Ca2+ influx that results in neuronal death. Mutations in NMDAR2B have been linked to early infantile epileptic encephalopathy that is characterized by refractory seizures and neurodevelopmental impairment. Some mutations are also known to cause mental retardation with or without seizures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2B
Applications Western Blot
Classification Monoclonal
Clone 11D1
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected NMDAR2B in Western Blotting applications (Lu, W., et. al. (2017). Science. 355(6331)). Western Blotting Analysis: 4μg/mL from a representative lot detected NMDAR2B in mouse brain tissue lysate.
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Glutamate receptor ionotropic NMDA 2B;GluN2B;Glutamate [NMDA] receptor subunit epsilon-2;N-methyl D-aspartate receptor subtype 2B;NR2B;N-methyl-D-aspartate receptor subunit 3;NR3;hNR3
Gene Symbols GRIN2B;NMDAR2B
Host Species Mouse
Immunogen Full length purified human Glutamate receptor ionotropic, NMDA 2B (NMDAR2B).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000825
Test Specificity Clone 11D1 is a mouse monoclonal antibody that specifically detects NMDAR2B.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.