Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2B, p Tyr1472 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB300182
Description
NMDAR2B Polyclonal specifically detects NMDAR2B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
NMDAR2B (p Tyr1472) | |
Polyclonal | |
Western Blot 1:1000, Simple Western | |
Q00960 | |
GRIN2B | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1472 conjugated to KLH. Accession # Q00960 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼180k NMDAR NR2B-subunit protein phosphorylated at Tyr1472 in Western blots. The antibody also labels proteins of ∼65k and ∼115k. Immunolabeling is completely blocked by blocked by either gamma-Ptase or by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
Rabbit | |
180 kDa | |
0.1 mL | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
2904 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction