Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2C Mouse anti-Rat, Unconjugated, Clone: N422/18, MilliporeSigma™

Catalog No. MABN18692MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN18692MI 25 μg
MABN18691MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN18692MI Supplier MilliporeSigma Supplier No. MABN186925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NMDAR2C Monoclonal antibody specifically detects NMDAR2C in Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2C
Applications Western Blot
Classification Monoclonal
Clone N422/18
Conjugate Unconjugated
Gene Accession No. NP_036707.3
Gene Alias Glutamate receptor ionotropic+C6 NMDA 2C;GluN2C;Glutamate [NMDA] receptor subunit epsilon-3;N-methyl D-aspartate receptor subtype 2C;NR2C
Gene Symbols Grin2c
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 398 amino acids from the C-terminal, cytoplasmic domain of rat Glutamate receptor ionotropic, NMDAR2C.
Molecular Weight of Antigen ∼150 kDa observed; 135.27 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone N422/18 is a mouse monoclonal antibody that specifically detects rat Glutamate receptor ionotropic, NMDAR2C. It targets an epitope within the C-terminal cytoplasmic domain.
Target Species Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.