Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541001 Supplier Invitrogen™ Supplier No. PA541001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VNTTNPSSLL TQICGLLGAA HVHGIVFEDN VDTEAVAQIL DFISSQTHVP Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 79%.

The NMDA Receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimers, epilepsy and ischemic neuronal cell death. The NMDA receptor is also one of the principal molecular targets for alcohol in the CNS. The NMDAR is also potentiated by protein phosphorylation. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of th NMDAR2 (NR2 A-D) subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GRIN2C
Gene Accession No. Q14957
Gene Alias GluN2C; GluRepsilon3; glutamate [NMDA] receptor subunit epsilon-3; glutamate ionotropic receptor NMDA type subunit 2C; glutamate receptor; glutamate receptor ionotropic, NMDA 2C; glutamate receptor ionotropic, NMDA 2C; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2C; glutamate receptor, ionotropic, NMDA2C; glutamate receptor, ionotropic, NMDA2C (epsilon 3); glutamate receptor, ionotropic, N-methyl D-aspartate 2C; Grin2c; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2C; NMDA glutamate receptor; NMDAR2C; N-methyl D-aspartate receptor subtype 2C; N-methyl-D-aspartate receptor subunit 2C; NR2; NR2C
Gene Symbols GRIN2C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GRIN2C (aa 69-118).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2905
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.