Learn More
Invitrogen™ NMDAR2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ OPA104020
Description
This antibody detects predominantly the NR2C isoform but may show low level cross-reactivity with the NR2A and NR2B isoforms. In Western blot, this antibody detects a ~140 kDa protein representing NR2C in rat brain homogenate. Faint bands representing the NR2A and NR2B isoforms at 180 KDa may also be observed. Immunohistochemical staining in rat brain with OPA1-04020 yields a pattern consistent with plasma membrane staining. Reconstitute with 50 μL of PBS.
The NMDA Receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimers, epilepsy and ischemic neuronal cell death. The NMDA receptor is also one of the principal molecular targets for alcohol in the CNS. The NMDAR is also potentiated by protein phosphorylation. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of th NMDAR2 (NR2 A-D) subunits.
Specifications
NMDAR2C | |
Polyclonal | |
Unconjugated | |
GRIN2C | |
GluN2C; GluRepsilon3; glutamate [NMDA] receptor subunit epsilon-3; glutamate ionotropic receptor NMDA type subunit 2C; glutamate receptor; glutamate receptor ionotropic, NMDA 2C; glutamate receptor ionotropic, NMDA 2C; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2C; glutamate receptor, ionotropic, NMDA2C; glutamate receptor, ionotropic, NMDA2C (epsilon 3); glutamate receptor, ionotropic, N-methyl D-aspartate 2C; Grin2c; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2C; NMDA glutamate receptor; NMDAR2C; N-methyl D-aspartate receptor subtype 2C; N-methyl-D-aspartate receptor subunit 2C; NR2; NR2C | |
Rabbit | |
Affinity chromatography | |
RUO | |
14813, 24411, 2905 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
5mM Ammonium bicarbonate with no preservative; pH 7.4 | |
Q00961, Q01098, Q14957 | |
GRIN2C | |
Fusion protein from the N-terminal region of the NR2C subunit of rat GRIN2C. | |
10 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.