Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR2D Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP100197
Description
NMDAR2D Polyclonal specifically detects NMDAR2D in Human, Mouse, Rat samples. It is validated for Peptide ELISA.Specifications
NMDAR2D | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
EB11estrogen receptor binding CpG island, glutamate receptor, ionotropic, N-methyl D-aspartate 2D, NMDA epsilon 4, NMDAR2DN-methyl-d-aspartate receptor subunit 2D, N-methyl D-aspartate receptor subtype 2D, NR2Dglutamate [NMDA] receptor subunit epsilon-4 | |
Goat | |
Affinity Purified | |
RUO | |
2906 | |
Store at -20C. Avoid freeze-thaw cycles. |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:32000 | |
NP_000827.2 | |
GRIN2D | |
Peptide with sequence C-TRRGSAHFSSLESE corresponding to C-Terminus according to NP_000827.2. | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction