Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2D Mouse anti-Rat, Clone: 1G9.39A5, MilliporeSigma™

Catalog No. MAB5578I25 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5578I25 25 μg
MAB5578I100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB5578I25 Supplier MilliporeSigma Supplier No. MAB5578I25UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NMDAR2D Monoclonal antibody specifically detects NMDAR2D in Rat samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2D
Applications Western Blot
Classification Monoclonal
Clone 1G9.39A5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Recombinant rat NMDAR2D.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.