Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101608 Supplier Invitrogen™ Supplier No. PA5101608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GRIN2D.

Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors exhibit slow kinetsis of Ca2+ ions and a high permeability for Ca2+ ions. The NMDA receptors consist of five subunits: epsilon 1, 2, 3, 4 and one zeta subunit. The zeta subunit is expressed throughout the brainstem whereas the four epsilon subunits display limited distribution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2D
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GRIN2D
Gene Accession No. O15399, Q03391, Q62645
Gene Alias EB11; estrogen receptor binding CpG island; GluN2D; glur epsilon 4; GluRepsilon4; glutamate [NMDA] receptor subunit epsilon-4; glutamate ionotropic receptor NMDA type subunit 2D; glutamate receptor; glutamate receptor ionotropic, NMDA 2D; glutamate receptor, ionotropic, NMDA2D; glutamate receptor, ionotropic, NMDA2D (epsilon 4); glutamate receptor, ionotropic, N-methyl D-aspartate 2D; GRIN2D; ionotropic NMDA glutamate receptor epsilon 4 subunit type 2; NMDAR2D; N-methyl D-aspartate receptor subtype 2D; N-methyl-d-aspartate receptor subunit 2D; NR2D
Gene Symbols GRIN2D
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRIN2D(Accession O15399), corresponding to amino acid residues Y673-R723.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14814, 24412, 2906
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.