Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR3A (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577431
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Grin3a is an NMDA (N-methyl-D-aspartate) receptor subtype of glutamate-gated ion channels with reduced single-channel conductance, low calcium permeability and low voltage-dependent sensitivity to magnesium. Grin3a is mediated by glycine and may play a role in the development of dendritic spines. Further, Grin3a may play a role in PPP2CB-NMDAR mediated signaling mechanism. Grin3a belongs to the superfamily of glutamate-regulated ion channels, and function in physiological and pathological processes in the central nervous system. Grin3a shows greater than 90% identity to the corresponding subunit in rat. Studies in the knockout mouse deficient in Grin3a suggest that the protein may be involved in the development of synaptic elements by modulating NMDA receptor activity. Diseases associated with GRIN3A include Lingual-Facial-Buccal Dyskinesia.
Specifications
NMDAR3A (extracellular) | |
Polyclonal | |
Unconjugated | |
Grin3a | |
GRIN3A, GluN3A, NMDAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
191573, 242443 | |
-20°C | |
Lyophilized |
Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q9R1M7 | |
Grin3a | |
Peptide (C)RHKTHFQHPNKLHLR, corresponding to amino acid residues 502-516 of rat NMDAR3A | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction