Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMDAR3B Monoclonal Antibody (2A8E11)
GREENER_CHOICE

Catalog No. PIMA531748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531748 Supplier Invitrogen™ Supplier No. MA531748
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31748 has been tested in indirect ELISA.

NMDA receptor subtype of glutamate-gated ion channels with reduced single-channel conductance, low calcium permeability and low voltage-dependent sensitivity to magnesium. Mediated by glycine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR3B
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2A8E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRIN3B
Gene Accession No. O60391
Gene Alias GluN3B; glutamate [NMDA] receptor subunit 3B; glutamate ionotropic receptor NMDA type subunit 3B; glutamate receptor; glutamate receptor ionotropic, NMDA 3B; glutamate receptor, ionotropic, NMDA3B; glutamate receptor, ionotropic, N-methyl-D-aspartate 3B; GRIN3B; NMDA receptor 4; NMDA receptor subunit 3B; NMDA type glutamate receptor subunit NR3B; NMDAR3B; N-methyl-D-aspartate receptor subtype 3B; N-methyl-D-aspartate receptor subunit NR3B; NR3B; Nr4
Gene Symbols GRIN3B
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIN3B (AA: 135-276) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.