Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                NME1 Monoclonal Antibody (OTI4H2), TrueMAB™, OriGene
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$516.29
Specifications
| Antigen | NME1 | 
|---|---|
| Clone | OTI4H2 | 
| Concentration | 1 mg/mL | 
| Applications | Immunohistochemistry (Paraffin), Western Blot | 
| Classification | Monoclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 50-167-8774    | OriGene TA801285 | 100 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $516.29
                                                 |  | |||||
Description
NME1 Monoclonal antibody specifically detects NME1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NME1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P15531 | |
| IgG2b | |
| -20° C, Avoid Freeze/Thaw Cycles | 
| OTI4H2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 4830 | |
| Primary | |
| NME1 | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
            